Learn More
R&D Systems™ Protein A ELISA Kit
Intended for detection of natural and recombinant Protein A in Bioprocess manufacturing applications
Marke:  R&D Systems™ BPPA00
Artikelnummer. 30000815
  • 848.00 CHF / 1 Set
Verfügbar ab: 17-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Spezifikation
46.90 to 3000 pg/mL
3919448
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
ELISA
1 Kit
Immunoglobulin G binding protein A, PROA, Protein-A, SPA, Staphylococcal protein A