Learn More
Thermo Scientific™ POROS™ XS RoboColumn™ Pre-packed columns
Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.
276.00 CHF - 358.00 CHF
Spezifikation
Stationäre Phase | Sulphopropyl Functional Group |
---|---|
Inhalt und Lagerung | Store at 2°C to 8°C or at room temperature. |
Produktlinie | POROS |
Versandbedingung | Raumtemperatur |
Aufreinigungsziel | Protein |
Produktcode | Marke | Volumen (metrisch) | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Volumen (metrisch) | Preis | Menge & Verfügbarkeit | |||||
18416418
|
Thermo Scientific™
A30715 |
200 μL |
276.00 CHF
200 Mikroliter |
Verfügbar ab: 25-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18476628
|
Thermo Scientific™
A30716 |
600 μL |
358.00 CHF
600 Mikroliter |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XS resin, a 50-micron strong cation-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS XS resin is the first high-capacity, high-resolution resin that allows loading to more than 100 mg/mL capacity in the presence of up to 15 mS/cm NaCl, while delivering unprecedented separation capability. A major improvement on traditional cation exchangers, POROS XS resin sets the standard for cation exchange chromatography.
Features of the resin:
- Dynamic binding capacity leads to reduced column size, smaller footprint, decreased buffer usage, and reduced cycling
- High resolution supports better product yield and improved impurity clearance
- Robust salt tolerance allows improved process flexibility, alignment, and process flow streamlining
- Low back pressure and linear pressure responses help improve scalability
- Rigid polymeric bead with covalent surface chemistry makes for easier handling and packing, and superior physical and chemical stability, resulting in a robust manufacturing process
RoboColumn™ is a registered trademark of ATOLL GmbH.
Spezifikation
Sulphopropyl Functional Group | |
POROS | |
Protein |
Store at 2°C to 8°C or at room temperature. | |
Raumtemperatur |