missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mutexil™ Polypropylene Hooded Jumpsuits
Click to view available options
Größe:
Large
Beschreibung
- Polypropylene non woven coverall with mao collar
- Elastics on ankle, wrist and back
- Zip fastening and two pockets
- Applications include maintenance, production site visits
- For use in non-hazardous environments
Spezifikation
Spezifikation
Produkttyp | General Purpose Coveralls |
Größe | Large |
Geschlecht | Unisex |
Material | Polypropylen |
Farbe | White |
Einwegprodukt | Single-use |
Taillenform | Elastisch |
Knöchelform | Elastisch |
Sterilität | Non-sterile |
Anzahl Taschen | 2 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'