missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platin-Standardlösung, Plasma HIQU, Chem Lab™
Click to view available options
Menge:
100 ml
Verpackung:
100 ml
Spezifikation
Konzentration | 1000 μg/ml |
Verpackung | 100 ml |
Menge | 100 ml |
Farbe | Braun |
Geruch | Geruchsneutral |
pH | <1 |
Physikalische Form | Flüssig |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'