missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Colour Reference Standart Pt-Co 5 500 ML
Click to view available options
Menge:
500 mL
Spezifikation
Menge | 500 mL |
Geruch | Odorless |
Lösung enthält | Hexachloroplatinate of Potassium, Cobalt chloride, Hydrochloric acid |
pH | ∼3 |
Löslichkeit | Miscible with water |
Physikalische Form | Solution |
Sicherheit und Handhabung
Produktidentifikator
- reference standard Pt-Co 30/ platinum cobalt 30
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'