missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRB (PDGFR β) Recombinant Human Protein
PDGFRB is a receptor tyrosine kinase.
Marke: Invitrogen™ PR4465B
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PDGFRB has been implicated in atherogenesis and in cell transformation by the SIS oncogene.
Spezifikation
NP_002600 | |
20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
PDGFRB (PDGFR β) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
66.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |