missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRA [D842V] Recombinant Human Protein
PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.
Marke: Invitrogen™ PV4292
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PDGFRA D842V is expressed in a high percentage of epithelial ovarian cancers and is implicated in the pathogenesis of acute myelogenous leukemia.
Spezifikation
NP_006197, D842V | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
PDGFRA D842V, PDGFR alpha | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
89.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |