Learn More
Tocris Bioscience™ PD 4'-piperazine
Stable cereblon ligand, for PROTAC synthesis
Marke: Tocris Bioscience™ 7859/50
308.42 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Beschreibung
PD 4'-piperazine is a cereblon ligand that can be used for PROTAC® Degrader development. The compound can be used to develop PD-PROTAC molecules that exhibit improved stability and potency compared with PROTACs synthesized using IMiD (immunomodulatory imide drug) analogs.Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Spezifikation
Dihydro-1-[4-(1-piperazinyl)phenyl]-2,4(1H,3H)-pyrimidinedione dihydrochloride | |
50 mg | |
Degrader Building Blocks | |
C14H18N4O2.2HCl |