missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Mouse anti-Human, Mouse, Porcine, Rat, Clone: 10D10E11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60097-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associates with formin-binding protein 17, but its function is unknown. This protein may form oligomeric complexes with family members.Spezifikation
PCNA | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04961, P12004, P17918 | |
PCNA | |
PCNA Fusion Protein Ag7416 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
10D10E11 | |
Unconjugated | |
PCNA | |
Cyclin, PCNA | |
Mouse | |
Protein G | |
RUO | |
18538, 25737, 5111, 692192 | |
-20°C | |
Liquid |