missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ APO-BRDU-IHC (TUNEL) Apoptosis Kit
Applications: Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Marke: Novus Biologicals™ NBP2-31164
Additional Details : Gewicht : 0.58200kg
Spezifikation
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Positive & Negative Control Slides, Blocking Buffer, H202/Urea Tablets, Proteinase K, DAB Tablets, TdT Enzyme, Br-dUTP, 200X Conjugate, 5X-Reaction Buffer, Biotin-anti BrdU antibody, Methyl Green | |
APO-BRDU-IHC (TUNEL) Apoptosis Kit |
1 Kit | |
Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin |