Learn More
Nichrome-Etzanchant, Thermo Scientific Chemicals
High Purity Ceric Ammonium Nitrate System Provides Clean, Precise Etching of Nichrome Layers For Preparation of Thin Film Circuits
89.15 CHF
Spezifikation
Chemischer Name oder Material | Nichrome etchant |
---|---|
Farbe | Farblos |
MDL-Nummer | MFCD07370976 |
Geruch | Acrid |
Form | Flüssig |
Beschreibung
High purity ceric ammonium nitrate system provides clean, precise etching of nichrome layers for preparation of thin film circuits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Nichrome etchant | |
MFCD07370976 | |
Flüssig |
Farblos | |
Acrid |
Sicherheit und Handhabung
When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P201-P202-P234-P260-P263-P264b-P270-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H331-H360D-H362
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Umgebungstemperaturen
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.