Learn More
NFE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11089-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This is a rabbit polyclonal antibody raised against full-length NFE2 of human origin.
Component of the NF-E2 complex essential for regulating erythroid and megakaryocytic maturation and differentiation. Binds to the hypersensitive site 2 (HS2) of the beta-globin control region (LCR). This subunit (NFE2) recognizes the TCAT/C sequence of the AP-1-like core palindrome present in a number of erythroid and megakaryocytic gene promoters. Requires MAFK or other small MAF proteins for binding to the NF-E2 motif. May play a role in all aspects of hemoglobin production from globin and heme synthesis to procurement of iron.Spezifikation
NFE2 | |
Polyclonal | |
Unconjugated | |
NFE2 | |
Leucine zipper protein NF E2, NF E2, NFE2, p45, p45 NF E2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4778 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16621 | |
NFE2 | |
NFE2 Fusion Protein Ag1570 | |
150 μL | |
Primary | |
Human | |
Antibody | |