Learn More
Invitrogen™ Neon™ NxT Electroporation System Pipette
The Neon NxT Electroporation System Pipette is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1).
Marke: Invitrogen™ NEON1P
Additional Details : Gewicht : 0.34000kg
Beschreibung
The Neon NxT Electroporation System Pipette is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1). Each system comes with a pipette. This pipette is offered separately for those needing an additional pipette or replacement for their existing pipette.
Spezifikation
Neon™ NxT tips (Cat. No. N1025, N10025, N1096, N10096) | |
Electroporation Pipette | |
Transfektion |
Neon NxT | |
10, 100 μL | |
Neon™ NxT Electroporation System |
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.