missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP1 / ABCC1 Mouse, Clone: MRP1/1343, enQuireBio™
Mouse Monoclonal Antibody
234.00 CHF - 484.00 CHF
Spezifikation
Antigen | MRP1 / ABCC1 |
---|---|
Klon | MRP1/1343 |
Konzentration | 200μg/mL |
Anwendungen | Immunohistochemistry |
Klassifikation | Monoclonal |
Spezifikation
MRP1 / ABCC1 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
4363 | |
Recombinant human full-length ABCC1 | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
190kDa |
MRP1/1343 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_016878731 | |
ABCC1 | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic and Cell Surface | |
A549 cells. Colon, Endometrium, Ovary. Testis or Tonsil. |