Learn More
R&D Systems™ Mouse TIM-1/KIM-1/HAVCR DuoSet ELISA
Sufficient for at least 15 96-well plates
Marke:  R&D Systems™ DY1817
Artikelnummer. 30021801
  • 854.00 CHF / 1 Set
Verfügbar ab: 11-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Mouse TIM-1/KIM-1/HAVCR DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TIM-1/KIM-1/HAVCR.
Spezifikation
Spezifikation
Biotin
ELISA
CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
31.20 to 2000 pg/mL
171283
Store the unopened product at 2°C to 8°C. Do not use past expiration date.