missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Autoimmune Regulator/AIRE Antibody, R&D Systems™
Goat Polyclonal Antibody
Marke: R&D Systems AF6184
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Autoimmune Regulator/AIRE Polyclonal specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Western Blot.Spezifikation
Autoimmune Regulator/AIRE | |
Polyclonal | |
Unconjugated | |
Q9Z0E3 | |
AIRE | |
E. coli-derived recombinant mouse Autoimmune Regulator/AIRE Ser476-Ser552 Accession # Q9Z0E3 | |
100 μg | |
Primary | |
Detects mouse Autoimmune Regulator/AIRE in Western blots. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AIRE, APECED, APECED protein, APECEDAPSI, APS1, APSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1 | |
Goat | |
Affinity Purified | |
RUO | |
326 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |