missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Autoimmune Regulator/AIRE Antibody, R&D Systems™
Rat Monoclonal Antibody
Marke: R&D Systems MAB6184-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Autoimmune Regulator/AIRE Monoclonal specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Spezifikation
Autoimmune Regulator/AIRE | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q9Z0E3 | |
AIRE | |
E. coli-derived recombinant mouse AIRE Ser476-Ser552 (predicted) Accession # Q9Z0E3 | |
25 μg | |
Primary | |
Detects mouse AIRE in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human AIRE is observed. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
609930 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AIRE, APECED, APECED protein, APECEDAPSI, APS1, APSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
326 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur