missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MET [M1250T] Recombinant Human Protein
MET [M1250T] Recombinant Human Protein
Marke: Invitrogen PV3968
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MET (cMET) is a receptor-like tyrosine kinase whose disregulation has been linked to many types of human malignancies. The M1250T mutant of MET is both activating and displays altered substrate specificity.Spezifikation
NP_000236.2, M1250T | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
53.7kDa | |
10 μg | |
For research use only. Not for use in diagnostic procedures | |
MET [M1250T] Recombinant Human Protein | |
MET (HGFR) | |
MET [M1250T] Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
0.31mg/mL | |
4233, 4790 | |
MET [M1250T] Recombinant Human Protein | |
–80°C | |
MET M1250T | |
MET M1250T was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 250μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 956-1390; M1250T | |
176nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
His Tag | |
Kinase and inhibitors | |
70% by SDS-PAGE |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only. Not for use in diagnostic procedures.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur