missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP tag Mouse anti-Tag, Clone: 4C6H4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66003-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The approach of epitope tagging offers an easy and universal strategy for the identification and purification of proteins derived by recombit DNA technology. The insertion of a Maltose Binding Protein (MBP) tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the MBP-tagged product. Cleavage by factor Xa separates MBP from its partner protein.Spezifikation
MBP tag | |
Monoclonal | |
1.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Mouse | |
Protein A | |
RUO | |
-20°C | |
Liquid | |
Tag |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
4C6H4 | |
Unconjugated | |
Maltose Binding Protein, MBP, MBP Tag | |
MBP tag Fusion Protein Ag0942 | |
150 μL | |
Primary | |
Antibody | |
IgG2a |