missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ MATRIX Orbital Delta F1.5 Thermoshaker
The MATRIX Orbital Delta F1.5 Thermoshaker mixes and heats with a fixed insert which holds 24 tubes (1.5 mL). Whether its blood samples, pharmaceutical agents, DNA/RNA samples, or ELISA assays, samples with even the smallest volumes mix reliably and completely in all laboratory applications.
2499.98 CHF
Spezifikation
Orbit | 3 mm |
---|---|
Tragkraft (metrisch) | 0.3 kg |
Genauigkeit der Drehzahlregelung | ±30 rpm |
Drehzahlbereich | 0 to 1500 rpm |
Spannung | 100/240 V |
Beschreibung
- Large and clear display with a convenient menu structure allows for intuitive operation
- Speeds of up to 1500 rpm and shaker diameter of 3 mm ensure the liquid mixes quickly and effectively
- Two different speed modes for gentle heating of the samples
- Programmable ramps allow for automatic execution of mixing and heating tasks
- Aluminum die cast housing ensures secure positioning, excellent stability, and general robustness
Spezifikation
3 mm | |
±30 rpm | |
100/240 V | |
240 W | |
CE | |
320 x 200 x 120 mm | |
Touch Key Interface | |
TFT LCD | |
Fixed Insert for 24 1.5 mL Tubes |
0.3 kg | |
0 to 1500 rpm | |
100°C (Max.) | |
Orbital Shaker with Heating | |
±0.5°C | |
9.2 kg | |
MATRIX Orbital Delta F1.5 | |
80% |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts