Learn More
Novus Biologicals™ Maltose Binding Protein Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Marke:  Novus Biologicals™ NBC1-18538
Artikelnummer. 18748583
  • 532.00 CHF / 0.50 Milligramm
Verfügbar ab: 21-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
An un-tagged recombinant protein corresponding to the amino acids 27-392 of E.coli Maltose Binding Protein The Maltose Binding Protein Recombinant Protein is derived from E. coli. The Maltose Binding Protein Recombinant Protein has been validated for the following applications: SDS-Page.
Spezifikation
Spezifikation
0.5 mg