missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltose Binding Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1989
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PA1-989 detects recombinant maltose binding protein. PA1-989 has been used successfully in Western blotting to detect the presence of maltose binding protein tagged proteins. The PA1-989 immunogen is recombinant maltose binding protein.
The approach of epitope tagging offers an easy and universal strategy for the identification and purification of proteins derived by recombinant DNA technology. The insertion of a Maltose Binding Protein (MBP) tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the MBP-tagged product. Cleavage by factor Xa separates MBP from its partner protein.
Spezifikation
Maltose Binding Protein | |
Polyclonal | |
Unconjugated | |
MBP; MBP Tag | |
Recombinant maltose binding protein | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Tag | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur