Learn More
Abnova™ MAGEA1 (Human) Recombinant Protein
Human MAGEA1 full-length ORF ( AAH17555, 1 a.a. - 309 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00004100-P01.25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Sequence: MSLEQRSLHCKPEEALEAQQEALGLVCVQAAASSSSPLVLGTLEEVPTAGSTDPPQSPQGASAFPTTINFTRQRQPSEGSSSREEEGPSTSCILESLFRAVITKKVADLVGFLLLKYRAREPVTKAEMLESVIKNYKHCFPEIFGKASESLQLVFGIDVKEADPTGHSYVLVTCLGLSYDGLLGDNQIMPKTGFLIIVLVMIAMEGGHAPEEEIWEELSVMEVYDGREHSAYGEPRKLLTQDLVQEKYLEYRQVPDSDPARYEFLWGPRALAETSYVKVLEYVIKVSARVRFFFPSLREAALREEEEGV
Spezifikation
AAH17555 | |
melanoma antigen family A, 1 (directs expression of antigen MZ2-E) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAGEA1 | |
GST |
4100 | |
Wheat germ expression system | |
25 μg | |
MAGE1, MGC9326 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |