missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LM11A 31 dihydrochloride
Nonpeptide p75NTR ligand; neuroprotective
182.00 CHF - 668.00 CHF
Spezifikation
Reinheit | 95% |
---|---|
Summenformel | (2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride |
CAS | 1243259-19-9 |
Inhalt und Lagerung | Desiccate at RT |
Produkttyp | LM11A 31 dihydrochloride |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18767048
|
Tocris Bioscience™
5046/50 |
50 mg |
668.00 CHF
50 Milligramm |
Verfügbar ab: 19-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18721468
|
Tocris Bioscience™
5046/10 |
10 mg |
182.00 CHF
10 Milligramm |
Verfügbar ab: 19-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Nonpeptide p75NTR ligand; blocks p75-mediated cell death and also increases proliferation and survival of hippocampal neural progenitors. Exhibits no effect on nerve growth factor (NGF) binding to TrkA. Prevents and reverses atrophy of cholinergic neurites, as well as reversing Alzheimer's Disease (AD) pathologies in mid- to late-stage AD mice models. Shown to promote functional recovery in a mouse model of spinal cord injury. Orally available and brain penetrant.Spezifikation
95% | |
1243259-19-9 | |
LM11A 31 dihydrochloride | |
LM11A 31 dihydrochloride |
(2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride | |
Desiccate at RT | |
M.W. (Observed): 316.27 |
Nur für Forschungszwecke