Learn More
Epredia™ Lab Vision™ Caldesmon, HMW, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Caldesmon, HMW Ab-1, Mouse Monoclonal Antibody.
179.00 CHF - 605.00 CHF
Spezifikation
Antigen | Caldesmon, HMW Ab-1 |
---|---|
Klon | h-CALD |
Anwendungen | Immunohistochemistry (Paraffin) |
Klassifikation | Monoclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Form | Konzentration | Menge | Preis | Menge & Verfügbarkeit | |||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Form | Konzentration | Menge | Preis | Menge & Verfügbarkeit | |||
12659186
|
Epredia™
MS-1169-P0 |
Concentrated | 200μg/mL | 100 μL |
179.00 CHF
100 Mikroliter |
Verfügbar ab: 17-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||
12679186
|
Epredia™
MS-1169-P1 |
Concentrated | 200μg/mL | 500 μL |
477.00 CHF
500 Mikroliter |
Verfügbar ab: 17-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||
12649186
|
Epredia™
MS-1169-P |
Concentrated | 200μg/mL | 1 mL |
605.00 CHF
1 Milliliter |
Verfügbar ab: 17-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||
Beschreibung
Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.Host Species: Mouse
Clone: h-CALD
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Crude human uterus extract
Molecular Weight: 150kDa (h-caldesmon)
Positive Control: Uterus
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
Caldesmon, HMW Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Crude human uterus extract |
h-CALD | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Primary |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.