missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences X100 DISPOSAFLEX SAEULEN SET 8ML .
Inexpensive, multipurpose column for analytical separations using aqueous buffers
Marke: DWK Life Sciences 420160-0000
Includes:
- 8mL capacity, 8mm I.D. x 200mm long column
- 9mL capacity reservoir
- Lower column fitting with luer outlet and a recess to hold a porous bed support or a glass wool plug
Beschreibung
- Ideal for extraction of cyclic nucleotides cAMP and cGMP from sample preparations using AG1-X8
- All components are comprised of disposable polypropylene to eliminate cross contamination of labeled compounds
Bestellinformation
- Shipped unassembled
- Overall length of assembled unit: 265mm; 200mm length of column can be trimmed if necessary
Spezifikation
Analytical separations using aqueous Buffers | |
Chromatography Column Set | |
200 mm | |
Polyethylene |
8 mm | |
8 mL | |
Polypropylen | |
9mL Polypropylene reservoir, lower column fitting with luer outlet, Polyethylene bed supports, On-off valves |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur