missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRAK3, GST Tag Recombinant Protein
This IRAK3 Recombinant Human Protein (full length) was expressed in insect cells.
570.00 CHF - 8240.00 CHF
Spezifikation
Zugriffsnummer | AAH57800 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Molekulargewicht | 94.0 kDa |
Kinase-Familie | IRAK Kinase Family |
Proteinlänge | Full Length |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15930303
|
Invitrogen™
A30975 |
10 μg |
570.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17128795
|
Invitrogen™
A33378 |
1 mg |
8240.00 CHF
1 Milligramm |
Verfügbar ab: 15-11-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15970383
|
Invitrogen™
A32893 |
100 μg |
2645.00 CHF
100 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Spezifikation
AAH57800 | |
94.0 kDa | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 50 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Baculovirus | |
Recombinant, Full Length |
Kinase Assay | |
IRAK Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |