Learn More
IgM Mouse, CoraLite Plus 555, Clone: MOPC-104E, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL555-65243-100UG
Additional Details : Gewicht : 0.00100kg
Beschreibung
The IgM Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgM Isotype Control. This antibody recognizes antigen. The IgM Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 555.
Spezifikation
IgM | |
MOPC-104E | |
CoraLite Plus 555 | |
Mouse | |
Primary | |
900 kDa | |
IgM λ |
Flow Cytometry | |
0.2 mg/mL | |
IgM, IgM Isotype Control, Mouse IgM Isotype Control | |
100 μg | |
Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |