Learn More
IgG2b Mouse, CoraLite 647, Clone: 11B8C4, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL647-66360-3-100UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The Mouse IgG2b isotype control antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of Mouse IgG2b isotype control. This antibody recognizes Megathura crenulata antigen. The Mouse IgG2b isotype control antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Spezifikation
IgG2b | |
11B8C4 | |
Mouse | |
Isotype Control (Primary) | |
IgG2b |
Flow Cytometry | |
CoraLite 647 | |
100 μL | |
M. crenulata |