Learn More
IgG2a Mouse, CoraLite Plus 594, Clone: MOPC-173, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-65244-100UG
Additional Details : Gewicht : 0.00100kg
Beschreibung
The IgG2a Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgG2a Isotype Control. This antibody recognizes antigen. The IgG2a Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 594.
Spezifikation
IgG2a | |
MOPC-173 | |
CoraLite Plus 594 | |
Mouse | |
Primary | |
155 kDa | |
IgG2a κ |
Flow Cytometry | |
0.2 mg/ml | |
IgG1, IgG1 Isotype Control, Mouse IgG1 Isotype Control | |
100 μg | |
Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |