Learn More
IgG1 Mouse, CoraLite 647, Clone: 1F8D3, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL647-66360-100UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The Mouse IgG1 isotype control antibody from Proteintech is a mouse monoclonal antibody generated with peptide of Mouse IgG1 isotype control. This antibody recognizes Megathura crenulata antigen. The Mouse IgG1 isotype control antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Spezifikation
IgG1 | |
1F8D3 | |
Mouse | |
Isotype Control (Primary) | |
IgG1 |
Flow Cytometry | |
CoraLite 647 | |
100 μL | |
M. crenulata |