Learn More
IgG1 Mouse, CoraLite 555, Clone: MOPC-21, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL555-65124-100UG
Additional Details : Gewicht : 0.00100kg
Beschreibung
The IgG1 Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgG1 Isotype Control. This antibody recognizes antigen. The IgG1 Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is coralite™555.
Spezifikation
IgG1 | |
MOPC-21 | |
CoraLite 555 | |
Mouse | |
Primary | |
Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |
Flow Cytometry | |
0.2 mg/ml | |
IgG1, IgG1 Isotype Control, mouse IgG1, Mouse IgG1 Isotype Control | |
100 μg | |
16017 | |
155 kDa | |
IgG1 |