Learn More
Proteintech™ HumanKine™ Recombinant Human PDGFaa Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Marke:  Proteintech™ HZ-1215-10UG
Additional Details : Gewicht : 0.00010kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16861366
  • 130.93 CHF / 10 Mikrogramm
Verfügbar ab: 31-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spezifikation
Spezifikation
5154
10 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 10 mM HOAc containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 10 ng/mL EC50
Lyophilized
35, 40 and 45 kDa, homodimer, glycosylated
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%