Learn More
Proteintech™ HumanKine™ Recombinant Human PDGFaa Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Brand: Proteintech™ HZ-1215-100UG
Additional Details : Gewicht : 0.00010kg
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
5154 | |
100 μg | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 10 mM HOAc containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Typically ≤ 10 ng/mL EC50 | |
Lyophilized |
35, 40 and 45 kDa, homodimer, glycosylated | |
Human | |
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 95% |