missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human BMP-7 Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
130.93 CHF - 3449.22 CHF
Spezifikation
Gen-ID (Entrez) | 655 |
---|---|
Molekulargewicht | 29 kDa, homodimer, glycosylated |
Spezies | Human |
Rekonstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in 10 mM acetic acid containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
Inhalt und Lagerung | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16851376
|
Proteintech™
HZ-1229-10UG |
10 μg |
130.93 CHF
10 Mikrogramm |
Verfügbar ab: 31-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16841376
|
Proteintech™
HZ-1229-100UG |
100 μg |
690.99 CHF
100 Mikrogramm |
Verfügbar ab: 31-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16831376
|
Proteintech™
HZ-1229-1000UG |
1000 μg |
3449.22 CHF
1000 Mikrogramm |
Verfügbar ab: 31-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spezifikation
655 | |
Human | |
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 95% |
29 kDa, homodimer, glycosylated | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in 10 mM acetic acid containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Typically ≤ 100 ng/mL EC50 | |
Lyophilized |