missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Transglutaminase 3/TGM3 ELISA Kit
ELISA
Brand: Invitrogen™ EH463RBX5
This item is not returnable.
View return policy
Description
Human Transglutaminase 3/TGM3 quantitates human Transglutaminase 3/TGM3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Transglutaminase 3/TGM3.
Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene consists of two polypeptide chains activated from a single precursor protein by proteolysis. The encoded protein is involved the later stages of cell envelope formation in the epidermis and hair follicle.
Specifications
AI893889; E polypeptide; E polypeptide, protein-glutamine-gamma-glutamyltransferase; MGC126249; MGC126250; protein-glutamine gamma-glutamyltransferase E; Protein-glutamine gamma-glutamyltransferase E 27 kDa non-catalytic chain; Protein-glutamine gamma-glutamyltransferase E 50 kDa catalytic chain; protein-glutamine-gamma-glutamyltransferase; RGD1561831; TG; TG E; TG(E); TGase E; TGase TGase-3; Tgase3; TGase-3; TGE; Tgm3; transglutaminase 3; transglutaminase 3, E polypeptide; transglutaminase E; Transglutaminase-3 | |
0.16 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
TGM3 | |
<10 | |
HRP | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
0.16 to 40 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
7053 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
TGM3 | |
4 hrs 45 mins |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human Transglutaminase 3/TGM3 ELISA Kit > 5 x 96 Tests; HRP
Spot an opportunity for improvement?Share a Content Correction