missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TIM-3 Quantikine ELISA Kit
For the quantitative determination of human T cell Immunoglobulin and Mucin domain 3 (TIM-3) concentrations in cell culture supernates, cell lysates, serum, plasma, and urine. Human TIM-3 Immunoassay employs the quantitative sandwich enzyme immunoassay technique.
Marke: R&D Systems™ DTIM30
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human TIM-3 Quantikine ELISA Kit quantifies human TIM-3 in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (10 ul).Spezifikation
31.30 to 2000 pg/mL | |
HRP | |
Cell Culture Supernatant, Zelllysate, Plasma (EDTA, Heparin), Serum, Urin | |
ELISA | |
HAVCR2 | |
Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (10uL) | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 |
8.75 pg/mL | |
<0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested | |
Natural and Recombinant Human TIM-3 | |
84868 | |
1 Kit | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
TIM-3 |