Learn More
R&D Systems™ Human TIM-3 DuoSet ELISA
Sufficient for at least 15 96-well plates
Marke: R&D Systems™ DY2365
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human TIM-3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TIM-3.Spezifikation
1 Kit (for 15 Plates) | |
Biotin | |
ELISA | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 |
Human | |
62.50 to 4000 pg/mL | |
84868 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |