Learn More
R&D Systems™ Human TIM-1/KIM-1/HAVCR DuoSet ELISA
Human TIM-1/KIM-1/HAVCR DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke: R&D Systems™ DY1750B
Includes: Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Beschreibung
The Human TIM-1/KIM-1/HAVCR DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TIM-1/KIM-1/HAVCR.Spezifikation
1 Kit (for 15 Plates) | |
Biotin | |
15.60 to 1000 pg/mL | |
26762 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Human | |
TIM-1/KIM-1/HAVCR | |
ELISA | |
CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |