missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Serum TIM-1/KIM-1/HAVCR Quantikine ELISA Kit
For the determination of natural and recombinant TIM-1 in humans. R&D Systems Human Serum TIM-1/KIM-1/HAVCR Quantikine™ ELISA Kit is a 4.5 hour solid-phase ELISA designed to measure TIM-1 in human cell culture supernates, serum and plasma.
Marke: R&D Systems™ DSKM100
Includes: NS0-expressed recombinant human TIM-1
Beschreibung
The Human Serum TIM-1/KIM-1/HAVCR Quantikine ELISA Kit quantifies human TIM-1/KIM-1/HAVCR in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).Spezifikation
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
1 Kit | |
3.63 pg/mL | |
<0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested | |
26762 | |
50 μL (Serum), 50 μL (Cell Culture Supernates), 50 μL (Heparin plasma), 50 μL (EDTA plasma) | |
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
ELISA-Kit | |
Human | |
10.90 to 700 pg/mL | |
ELISA | |
Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL) | |
4.5 hr. | |
NS0-expressed recombinant human TIM-1 |