missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RBM34 Full-length ORF (NP_055829.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00023029-P01.10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Sequence: MSKRKRKRSVQEGENPDDGVRGSPPEDYRLGQVASSLFRGEHHSRGGTGRLASLFSSLEPQIQPVYVPVPKQTIKKTKRNEEEESTSQIERPLSQEPAKKVKAKKKHTNAEKKLADRESALASADLEEEIHQKQGQKRKNSQPGVKVADRKILDDTEDTVVSQRKKIQINQEEERLKNERTVFVGNLPVTCNKKKLKSFFKEYGQIESVRFRSLIPAEGTLSKKLAAIKRKIHPDQKNINAYVVFKEESAATQALKRNGAQIADGFRIRVDLASETSSRDKRSVFVGNLPYKVEESAIEKHFLDCGSIMAVRIVRDKMTGIGKGFGYVLFENTDSVHLALKLNNSELMGRKLRVMRSVNKEKFKQQNSNPRLKNVSKPKQGLNFTSKTAEGHPKSLFIGEKAVLLKTKKKGQKKSGRPKKQRKQKSpezifikation
NP_055829.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0117 | |
RBM34 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23029 | |
RBM34 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSKRKRKRSVQEGENPDDGVRGSPPEDYRLGQVASSLFRGEHHSRGGTGRLASLFSSLEPQIQPVYVPVPKQTIKKTKRNEEEESTSQIERPLSQEPAKKVKAKKKHTNAEKKLADRESALASADLEEEIHQKQGQKRKNSQPGVKVADRKILDDTEDTVVSQRKKIQINQEEERLKNERTVFVGNLPVTCNKKKLKSFFKEYGQIESVRFRSLIPAEGTLSKKLAAIKRKIHPDQKNINAYVVFKEESAATQALKRNGAQIADGFRIRVDLASETSSRDKRSVFVGNLPYKVEESAIEKHFLDCGSIMAVRIVRDKMTGIGKGFGYVLFENTDSVHLALKLNNSELMGRKLRVMRSVNKEKFKQQNSNPRLKNVSKPKQGLNFTSKTAEGHPKSLFIGEKAVLLKTKKKGQKKSGRPKKQRKQK | |
RUO | |
RBM34 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |