missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 2/CD18 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Marke: R&D Systems AF1730
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Integrin beta 2/CD18 Polyclonal specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Flow Cytometry, Adhesion Blockade, Immunocytochemistry, CyTOF-ready.Spezifikation
Integrin beta 2/CD18 | |
Polyclonal | |
Unconjugated | |
AAA59490 | |
ITGB2 | |
Mouse myeloma cell line NS0-derived recombinant human Integrin beta 2 Gln23-Asn700 Accession # AAA59490 | |
100 μg | |
Primary | |
Detects human Integrin beta 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Integrin beta 1, rhIntegrin beta 3, and recombinant mouse Integrin beta 6 is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Adhesion Blockade, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
959 beta subunit precursor)10, CD18, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), ITGB2, LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, LFA-1, LFA-1 beta, MAC-1 beta, MF17, MFI7, P150, p150,95 beta | |
Goat | |
Affinity Purified | |
RUO | |
3689 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur