Learn More
R&D Systems™ Human HGFR/c-MET DuoSet ELISA
Human HGF R/c-MET DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke: R&D Systems™ DY358
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human HGF R/c-MET DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human HGFR/c-MET.Spezifikation
Biotin | |
ELISA | |
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met |
62.50 to 4000 pg/mL | |
4233 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |