missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GJA4 Full-length ORF (NP_002051.2, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002701-P01.25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Mutations in this gene have been associated with atherosclerosis and a higher risk of myocardial infarction. [provided by RefSeq]
Sequence: MGDWGFLEKLLDQVQEHSTVVGKIWLTVLFIFRILILGLAGESVWGDEQSDFECNTAQPGCTNVCYDQAFPISHIRYWVLQFLFVSTPTLVYLGHVIYLSRREERLRQKEGELRALPAKDPQVERALAAVERQMAKISVAEDGRLRIRGALMGTYVASVLCKSVLEAGFLYGQWRLYGWTMEPVFVCQRAPCPYLVDCFVSRPTEKTIFIIFMLVVGLISLVLNLLELVHLLCRCLSRGMRARQGQDAPPTQGTSSDPYTDQVFFYLPVGQGPSSPPCPTYNGLSSSEQNWANLTTEERLASSRPPLFLDPPPQNGQKPPSRPSSSASKKQYVSpezifikation
NP_002051.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CX37 | |
GJA4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2701 | |
GJA4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGDWGFLEKLLDQVQEHSTVVGKIWLTVLFIFRILILGLAGESVWGDEQSDFECNTAQPGCTNVCYDQAFPISHIRYWVLQFLFVSTPTLVYLGHVIYLSRREERLRQKEGELRALPAKDPQVERALAAVERQMAKISVAEDGRLRIRGALMGTYVASVLCKSVLEAGFLYGQWRLYGWTMEPVFVCQRAPCPYLVDCFVSRPTEKTIFIIFMLVVGLISLVLNLLELVHLLCRCLSRGMRARQGQDAPPTQGTSSDPYTDQVFFYLPVGQGPSSPPCPTYNGLSSSEQNWANLTTEERLASSRPPLFLDPPPQNGQKPPSRPSSSASKKQYV | |
RUO | |
GJA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |