missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Galectin-9 DuoSet ELISA
Human Galectin-9 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke: R&D Systems™ DY2045
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human Galectin-9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Galectin-9.Spezifikation
1 Kit (for 15 Plates) | |
Biotin | |
ELISA | |
ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein |
Human | |
93.80 to 6000 pg/mL | |
3965 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |