Learn More
R&D Systems™ Human FAAH (NP_001432.2) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Marke: R&D Systems™ RDC3457
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
FAAH | |
2166 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_001432.2 | |
Anandamide amidohydrolase 1, EC 3.5.1, EC 3.5.1.99, EC 3.5.1.n2, FAAH1, FAAH-1, fatty acid amide hydrolase, fatty-acid amide hydrolase 1, MGC102823, MGC138146, Oleamide hydrolase 1, PSAB | |
Human |