missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DACT1 Partial ORF (NP_057735.2, 738 a.a. - 836 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00051339-Q01.25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Sequence: VVDTSEDEQSNYTTNCFGDSESSVSEGEFVGESTTTSDSEESGGLIWSQFVQTLPIQTVTAPDLHNHPAKTFVKIKASHNLKKKILRFRSGSLKLMTTVSpezifikation
NP_057735.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVDTSEDEQSNYTTNCFGDSESSVSEGEFVGESTTTSDSEESGGLIWSQFVQTLPIQTVTAPDLHNHPAKTFVKIKASHNLKKKILRFRSGSLKLMTTV | |
RUO | |
DACT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51339 | |
DACT1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAPPER/DAPPER1/DPR1/FRODO/HDPR1/THYEX3 | |
DACT1 | |
Recombinant | |
wheat germ expression system |