missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CLEC18B (NP_001011880) VersaClone cDNA
C-type Lectin Domain Family 18, Member B
Marke: R&D Systems™ RDC1774
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human CLEC18B (NP_001011880) VersaClone cDNA clone contains the complete ORF for CLEC18B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CLEC18B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Bestellinformation
Ships at ambient temperature
Spezifikation
cDNA | |
NP_001011880 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
497190 (Human), 353287 (Mouse), 307851 (Rat) | |
Human |
hCLEC18B | |
10 μg at 0.2 μg/μL | |
1381bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |