missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CEACAM-1/CD66a DuoSet ELISA
Human CEACAM-1/CD66a DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke: R&D Systems™ DY2244
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human CEACAM-1/CD66a DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CEACAM1/CD66a.Spezifikation
1 Kit (for 15 Plates) | |
Biotin | |
ELISA | |
antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR |
Human | |
93.80 to 6000 pg/mL | |
634 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |