Learn More
Invitrogen™ Human Apoptosis-Enhancing Nuclease (aa 16-92) Control Fragment Recombinant Protein
Recombinant Protein
Marke:  Invitrogen™ RP98170
Artikelnummer. 30182189
  • 248.00 CHF / 100 Mikroliter
Verfügbar ab: 17-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Spezifikation
Q8WTP8
Blocking Assay, Control
64782
100 μL
RUO
AEN
Human
SLTIPNAKDVLRKRHKRRSRQHQRFMARKALLQEQGLLSMPPEPGSSPLPTPFGAATATEAASSGKQCLRAGSGSAP
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Apoptosis-Enhancing Nuclease
-20° C, Avoid Freeze/Thaw Cycles
2700083B06Rik; AEN; aen {ECO:0000250; apoptosis enhancing nuclease; apoptosis-enhancing nuclease; apoptosis-enhancing nuclease {ECO:0000250; interferon stimulated exonuclease gene 20kDa-like 1; interferon stimulated exonuclease gene 20-like 1; interferon-stimulated 20 kDa exonuclease-like 1; interferon-stimulated 20 kDa exonuclease-like 1 {ECO:0000312; ISG20L1; pp12744; RGD:1305051}; RGD1305051; SBBI58; UniProtKB:Q8WTP8}
Unconjugated
Recombinant
E. coli