Learn More
Invitrogen™ Human Apoptosis-Enhancing Nuclease (aa 16-92) Control Fragment Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP98170
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Spezifikation
Q8WTP8 | |
Blocking Assay, Control | |
64782 | |
100 μL | |
RUO | |
AEN | |
Human | |
SLTIPNAKDVLRKRHKRRSRQHQRFMARKALLQEQGLLSMPPEPGSSPLPTPFGAATATEAASSGKQCLRAGSGSAP | |
Liquid |
≥5.0 mg/mL | |
1M urea, PBS with no preservative; pH 7.4 | |
Apoptosis-Enhancing Nuclease | |
-20° C, Avoid Freeze/Thaw Cycles | |
2700083B06Rik; AEN; aen {ECO:0000250; apoptosis enhancing nuclease; apoptosis-enhancing nuclease; apoptosis-enhancing nuclease {ECO:0000250; interferon stimulated exonuclease gene 20kDa-like 1; interferon stimulated exonuclease gene 20-like 1; interferon-stimulated 20 kDa exonuclease-like 1; interferon-stimulated 20 kDa exonuclease-like 1 {ECO:0000312; ISG20L1; pp12744; RGD:1305051}; RGD1305051; SBBI58; UniProtKB:Q8WTP8} | |
Unconjugated | |
Recombinant | |
E. coli |